Learn More
Meso Scale Discovery U-PLEX Human CA1 ASYQP
SDP
Catalog No. 5000079154
Encompass
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The assay requires 50 L or less sample volume and is compatible with Meso Scale Discovery platforms It includes a 96-well plate with capture and detection antibodies calibrators and controls for standard curve generation and assay validation This assay efficiently quantifies CA1 important in cancer and inflammatory diseases
Catalog No. 50-000-79154 Supplier Meso Scale Discovery Supplier No. K151J9K22
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The assay requires 50 L or less sample volume and is compatible with Meso Scale Discovery platforms It includes a 96-well plate with capture and detection antibodies calibrators and controls for standard curve generation and assay validation This assay efficiently quantifies CA1 important in cancer and inflammatory diseases