Learn More
Meso Scale Discovery U-PLEX Human CD20 ASY 25 PL
SDP
Catalog No. 5000082035
Encompass
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 MESO QuickPlex SQ 120 QuickPlex SQ 120 Plate Reader and V-PLEX Plate Reader The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and Calibrator
Catalog No. 50-000-82035 Supplier Meso Scale Discovery Supplier No. K151F7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 MESO QuickPlex SQ 120 QuickPlex SQ 120 Plate Reader and V-PLEX Plate Reader The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and Calibrator