Learn More
Meso Scale Discovery U-PLEX Human CD20 ASY
SDP
Catalog No. 5000078772
Encompass
The U-PLEX Human CD20 Assay from Meso Scale Discovery is a multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay allows for the quantitative measurement of CD20 a membrane protein expressed on B cells which can aid in understanding immune system function and diagnosing B cell malignancies It requires a small sample volume and provides results in as little as 3 5 hours using the MESO QuickPlex SQ 120 instrument With minimal hands-on time required this assay enables efficient and accurate CD20 quantification
Catalog No. 50-000-78772 Supplier Meso Scale Discovery Supplier No. K151F7K1
The U-PLEX Human CD20 Assay from Meso Scale Discovery is a multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay allows for the quantitative measurement of CD20 a membrane protein expressed on B cells which can aid in understanding immune system function and diagnosing B cell malignancies It requires a small sample volume and provides results in as little as 3 5 hours using the MESO QuickPlex SQ 120 instrument With minimal hands-on time required this assay enables efficient and accurate CD20 quantification