Learn More
Meso Scale Discovery U-PLEX Human CD20 ASY
SDP
Catalog No. 5000081643
Encompass
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay that enables simultaneous quantification of CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls It allows sensitive reproducible and accurate measurement of CD20 levels for life science research applications With a testing time of just 3 5 hours this kit offers a rapid and efficient workflow
Catalog No. 50-000-81643 Supplier Meso Scale Discovery Supplier No. K151F7K2
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay that enables simultaneous quantification of CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls It allows sensitive reproducible and accurate measurement of CD20 levels for life science research applications With a testing time of just 3 5 hours this kit offers a rapid and efficient workflow