Learn More
Meso Scale Discovery U-PLEX Human CD20 ASY384-well
SDP
Catalog No. 5000078637
Encompass
The U-PLEX Human CD20 384-well Assay Kit from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a CD20 plate detection antibody read buffer sample diluent and calibrators to quantitatively measure CD20 levels It is suitable for studies of B cell development signaling activation proliferation and more With a streamlined workflow and small sample volume requirements this kit enables efficient profiling of CD20 biomarkers
Catalog No. 50-000-78637 Supplier Meso Scale Discovery Supplier No. K251F7K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human CD20 384-well Assay Kit from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a CD20 plate detection antibody read buffer sample diluent and calibrators to quantitatively measure CD20 levels It is suitable for studies of B cell development signaling activation proliferation and more With a streamlined workflow and small sample volume requirements this kit enables efficient profiling of CD20 biomarkers