Learn More
Meso Scale Discovery U-PLEX Human CD27 ASY 25 PL
SDP
Catalog No. 5000082465
Encompass
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for measuring CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with MESO QuickPlex SQ 120 and V-PLEX Plus instruments The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve allowing simultaneous measurement of CD27 and other biomarkers
Catalog No. 50-000-82465 Supplier Meso Scale Discovery Supplier No. K151G7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for measuring CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with MESO QuickPlex SQ 120 and V-PLEX Plus instruments The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve allowing simultaneous measurement of CD27 and other biomarkers