Learn More
Meso Scale Discovery U-PLEX Human CD27 ASY
SDP
Catalog No. 5000082008
Encompass
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for quantifying CD27 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It is ideal for monitoring CD27 in immune studies providing rapid and reliable results with minimal hands-on time
Catalog No. 50-000-82008 Supplier Meso Scale Discovery Supplier No. K151G7K2
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for quantifying CD27 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It is ideal for monitoring CD27 in immune studies providing rapid and reliable results with minimal hands-on time