Learn More
Meso Scale Discovery U-PLEX Human CD27 ASY384-well
SDP
Catalog No. 5000079407
Encompass
The Meso Scale Discovery U-PLEX Human CD27 384-Well Assay Kit is a multiplex immunoassay that measures CD27 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of up to 80 samples The assay has a detection range of 50-10 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator and control solutions It enables quantification of CD27 a member of the tumor necrosis factor receptor superfamily involved in regulation of B- and T-cell activation
Catalog No. 50-000-79407 Supplier Meso Scale Discovery Supplier No. K251G7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD27 384-Well Assay Kit is a multiplex immunoassay that measures CD27 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of up to 80 samples The assay has a detection range of 50-10 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator and control solutions It enables quantification of CD27 a member of the tumor necrosis factor receptor superfamily involved in regulation of B- and T-cell activation