Learn More
Meso Scale Discovery U-PLEX Human CD27 ASYQP
SDP
Catalog No. 5000078504
Encompass
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for measuring CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve enabling simultaneous CD27 and biomarker quantification It provides a sensitive reproducible method for CD27 measurement
Catalog No. 50-000-78504 Supplier Meso Scale Discovery Supplier No. K151G7K22
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for measuring CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve enabling simultaneous CD27 and biomarker quantification It provides a sensitive reproducible method for CD27 measurement