Learn More
Meso Scale Discovery U-PLEX Human CD27 ASYQP
SDP
Catalog No. 5000081986
Encompass
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for measuring CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with MESO QuickPlex SQ 120 instruments The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve It provides high specificity sensitivity and reproducibility for CD27 measurement
Catalog No. 50-000-81986 Supplier Meso Scale Discovery Supplier No. K151G7K21
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay for measuring CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use Requiring 50 L of sample it is compatible with MESO QuickPlex SQ 120 instruments The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve It provides high specificity sensitivity and reproducibility for CD27 measurement