Learn More
Meso Scale Discovery U-PLEX Human CD28 ASY384-well
SDP
Catalog No. 5000080725
Encompass
The U-PLEX Human CD28 384-well Assay Kit from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD28 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This assay kit includes a CD28-specific antibody coated plate detection antibody read buffer sample diluent calibrator and control It can test up to 80 samples in 384-well format with minimal sample volume requirement The kit is for research use only
Catalog No. 50-000-80725 Supplier Meso Scale Discovery Supplier No. K251H7K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human CD28 384-well Assay Kit from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD28 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra-/inter-assay precision This assay kit includes a CD28-specific antibody coated plate detection antibody read buffer sample diluent calibrator and control It can test up to 80 samples in 384-well format with minimal sample volume requirement The kit is for research use only