Learn More
Meso Scale Discovery U-PLEX Human CD28 ASYQP
SDP
Catalog No. 5000080794
Encompass
The U-PLEX Human CD28 Assay from Meso Scale Discovery is a multiplex immunoassay kit for measuring human CD28 in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and rapid results requiring 50 L or less sample volume Compatible with MSD instruments it includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation This assay efficiently analyzes CD28 crucial for T cell activation
Catalog No. 50-000-80794 Supplier Meso Scale Discovery Supplier No. K151H7K22
The U-PLEX Human CD28 Assay from Meso Scale Discovery is a multiplex immunoassay kit for measuring human CD28 in various biological samples It uses electrochemiluminescence (ECL) detection for high sensitivity and rapid results requiring 50 L or less sample volume Compatible with MSD instruments it includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation This assay efficiently analyzes CD28 crucial for T cell activation