Learn More
Meso Scale Discovery U-PLEX Human CD28 ASYQP
SDP
Catalog No. 5000081150
Encompass
The U-PLEX Human CD28 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CD28 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows simultaneous measurement of CD28 levels along with other biomarkers in one well This assay has high specificity and sensitivity with minimal cross-reactivity
Catalog No. 50-000-81150 Supplier Meso Scale Discovery Supplier No. K151H7K21
The U-PLEX Human CD28 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures CD28 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows simultaneous measurement of CD28 levels along with other biomarkers in one well This assay has high specificity and sensitivity with minimal cross-reactivity