Learn More
Meso Scale Discovery U-PLEX Human Clusterin ASY
SDP
Catalog No. 5000081638
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay requires a low sample volume of 50 L and provides results in under 5 hours It can be used to study clusterin levels in cancer neurodegenerative diseases and other research applications involving this multifunctional protein
Catalog No. 50-000-81638 Supplier Meso Scale Discovery Supplier No. K151B9K2
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay requires a low sample volume of 50 L and provides results in under 5 hours It can be used to study clusterin levels in cancer neurodegenerative diseases and other research applications involving this multifunctional protein