Learn More
Meso Scale Discovery U-PLEX Human Clusterin ASY
SDP
Catalog No. 5000081952
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is sensitive reproducible and provides results in just 3 5 hours The assay has a lower limit of detection of 0 03 ng/mL and an upper limit of 10 000 ng/mL It requires a low sample volume of 50 L The assay can be used to study Clusterin levels in cancer neurodegenerative diseases kidney diseases and for biomarker research
Catalog No. 50-000-81952 Supplier Meso Scale Discovery Supplier No. K151B9K1
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is sensitive reproducible and provides results in just 3 5 hours The assay has a lower limit of detection of 0 03 ng/mL and an upper limit of 10 000 ng/mL It requires a low sample volume of 50 L The assay can be used to study Clusterin levels in cancer neurodegenerative diseases kidney diseases and for biomarker research