Learn More
Meso Scale Discovery U-PLEX Human Clusterin ASYQP
SDP
Catalog No. 5000081637
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay can be run on MSD s SECTOR Imager 6000 for high throughput and minimal sample consumption Overall the U-PLEX Human Clusterin Assay enables reliable quantification of Clusterin an important biomarker from human samples
Catalog No. 50-000-81637 Supplier Meso Scale Discovery Supplier No. K151B9K22
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay can be run on MSD s SECTOR Imager 6000 for high throughput and minimal sample consumption Overall the U-PLEX Human Clusterin Assay enables reliable quantification of Clusterin an important biomarker from human samples