Learn More
Meso Scale Discovery U-PLEX Human Clusterin ASYQP
SDP
Catalog No. 5000081639
Encompass
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay can be run on MSD s SECTOR Imager platform and takes approximately 3 5 hours to complete It is intended for research use only
Catalog No. 50-000-81639 Supplier Meso Scale Discovery Supplier No. K151B9K21
The Meso Scale Discovery U-PLEX Human Clusterin Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Clusterin in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is provided in a convenient all-in-one kit has a detection range of 50-10 000 pg/mL and exhibits high sensitivity specificity and reproducibility The assay can be run on MSD s SECTOR Imager platform and takes approximately 3 5 hours to complete It is intended for research use only