Learn More
Meso Scale Discovery U-PLEX Human Complement C9 ASY
SDP
Catalog No. 5000079675
Encompass
The Meso Scale Discovery U-PLEX Human Complement C9 Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent It has a sensitivity of 0 04 ng/mL and an upper limit of 2500 ng/mL allowing high-throughput quantification with low sample volume
Catalog No. 50-000-79675 Supplier Meso Scale Discovery Supplier No. K151U9K2
The Meso Scale Discovery U-PLEX Human Complement C9 Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent It has a sensitivity of 0 04 ng/mL and an upper limit of 2500 ng/mL allowing high-throughput quantification with low sample volume