Learn More
Meso Scale Discovery U-PLEX Human Complement C9 ASY
SDP
Catalog No. 5000080506
Encompass
The Meso Scale Discovery U-PLEX Human Complement C9 Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a dynamic range of 78 - 50 000 pg/mL offering a robust tool for immunology research
Catalog No. 50-000-80506 Supplier Meso Scale Discovery Supplier No. K151U9K1
The Meso Scale Discovery U-PLEX Human Complement C9 Assay Kit quantifies Complement C9 in human serum plasma urine cell culture supernatants and other samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a dynamic range of 78 - 50 000 pg/mL offering a robust tool for immunology research