Learn More
Meso Scale Discovery U-PLEX Human CRP ASY 25 PL
SDP
Catalog No. 5000082234
Encompass
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate CRP quantification with a broad dynamic range The assay has high sensitivity with a lower limit of detection of 0 02 ng/mL and requires as little as 25 L of sample The U-PLEX Human CRP Assay provides a simple rapid and robust method for measuring CRP levels
Catalog No. 50-000-82234 Supplier Meso Scale Discovery Supplier No. K151L9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate CRP quantification with a broad dynamic range The assay has high sensitivity with a lower limit of detection of 0 02 ng/mL and requires as little as 25 L of sample The U-PLEX Human CRP Assay provides a simple rapid and robust method for measuring CRP levels