Learn More
Meso Scale Discovery U-PLEX Human CRP ASY
SDP
Catalog No. 5000081176
Encompass
The Meso Scale Discovery U-PLEX Human CRP Assay Kit is a multiplex immunoassay that measures C-reactive protein (CRP) levels in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG 10X Read Buffer Sample Diluent and CRP calibrators The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81176 Supplier Meso Scale Discovery Supplier No. K151L9K2
The Meso Scale Discovery U-PLEX Human CRP Assay Kit is a multiplex immunoassay that measures C-reactive protein (CRP) levels in human serum plasma and cell culture supernatant samples The assay utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG 10X Read Buffer Sample Diluent and CRP calibrators The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL