Learn More
Meso Scale Discovery U-PLEX Human CRP ASY
SDP
Catalog No. 5000082276
Encompass
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible method for detecting CRP levels between 0 04 - 2500 ng/mL This assay can be used to aid in evaluation of inflammatory conditions and infections The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and SULFO-TAG labeled detection antibody
Catalog No. 50-000-82276 Supplier Meso Scale Discovery Supplier No. K151L9K1
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible method for detecting CRP levels between 0 04 - 2500 ng/mL This assay can be used to aid in evaluation of inflammatory conditions and infections The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and SULFO-TAG labeled detection antibody