Learn More
Meso Scale Discovery U-PLEX Human CRP ASY384-well
SDP
Catalog No. 5000080696
Encompass
The Meso Scale Discovery U-PLEX Human CRP 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatants This 384-well multiplex assay kit includes a CRP-specific antibody coated plate and calibrator diluent that allows for the accurate quantitation of CRP across a broad dynamic range of 78 pg/mL to 50 000 pg/mL The assay utilizes MSD s proprietary SULFO-TAG labels that emit light upon electrochemical stimulation for high sensitivity detection This allows for the rapid and efficient measurement of CRP from small sample volumes
Catalog No. 50-000-80696 Supplier Meso Scale Discovery Supplier No. K251L9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CRP 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatants This 384-well multiplex assay kit includes a CRP-specific antibody coated plate and calibrator diluent that allows for the accurate quantitation of CRP across a broad dynamic range of 78 pg/mL to 50 000 pg/mL The assay utilizes MSD s proprietary SULFO-TAG labels that emit light upon electrochemical stimulation for high sensitivity detection This allows for the rapid and efficient measurement of CRP from small sample volumes