Learn More
Meso Scale Discovery U-PLEX Human CRP ASYQP 25 PL
SDP
Catalog No. 5000079298
Encompass
The Meso Scale Discovery U-PLEX Human CRP Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible method for detecting CRP levels between 0 3-2500 pg/mL This assay kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The U-PLEX platform provides a simple rapid workflow that delivers results in just 3 5 hours
Catalog No. 50-000-79298 Supplier Meso Scale Discovery Supplier No. K151L9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CRP Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible method for detecting CRP levels between 0 3-2500 pg/mL This assay kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and sample diluent The U-PLEX platform provides a simple rapid workflow that delivers results in just 3 5 hours