Learn More
Meso Scale Discovery U-PLEX Human CRP ASYQP
SDP
Catalog No. 5000079959
Encompass
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate CRP quantification with a broad dynamic range The assay requires a small sample volume and has a rapid 2 5 hour protocol The U-PLEX Human CRP Assay provides a simple fast and reliable tool for investigating CRP levels in research and clinical applications
Catalog No. 50-000-79959 Supplier Meso Scale Discovery Supplier No. K151L9K22
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate CRP quantification with a broad dynamic range The assay requires a small sample volume and has a rapid 2 5 hour protocol The U-PLEX Human CRP Assay provides a simple fast and reliable tool for investigating CRP levels in research and clinical applications