Learn More
Meso Scale Discovery U-PLEX Human CRP ASYQP
SDP
Catalog No. 5000079961
Encompass
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a sensitive and reproducible method for measuring CRP levels between 0 3-2500 pg/mL The assay can be run on MSD s SECTOR Imager instruments and provides results in as little as 3 5 hours The U-PLEX format allows for customization and multiplexing with other U-PLEX biomarker assays Overall the assay provides a rapid sensitive and flexible method for quantifying CRP levels in human samples
Catalog No. 50-000-79961 Supplier Meso Scale Discovery Supplier No. K151L9K21
The Meso Scale Discovery U-PLEX Human CRP Assay is an electrochemiluminescent immunoassay for the quantitative measurement of C-reactive protein (CRP) in human serum plasma and cell culture supernatant The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide a sensitive and reproducible method for measuring CRP levels between 0 3-2500 pg/mL The assay can be run on MSD s SECTOR Imager instruments and provides results in as little as 3 5 hours The U-PLEX format allows for customization and multiplexing with other U-PLEX biomarker assays Overall the assay provides a rapid sensitive and flexible method for quantifying CRP levels in human samples