Learn More
Meso Scale Discovery U-PLEX Human CTACK ASY 25 PL
SDP
Catalog No. 5000081933
Encompass
The Meso Scale Discovery U-PLEX Human CTACK Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of CTACK in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can perform up to 80 tests across two plates The lower limit of detection is 0 03 pg/mL The kit includes a 96-well MULTI-ARRAY Human CTACK Plate Diluent 100 Read Buffer T (4X) and vials of detection antibodies
Catalog No. 50-000-81933 Supplier Meso Scale Discovery Supplier No. K151VDK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CTACK Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of CTACK in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can perform up to 80 tests across two plates The lower limit of detection is 0 03 pg/mL The kit includes a 96-well MULTI-ARRAY Human CTACK Plate Diluent 100 Read Buffer T (4X) and vials of detection antibodies