Learn More
Meso Scale Discovery U-PLEX Human CTACK ASY
SDP
Catalog No. 5000079829
Encompass
The Meso Scale Discovery U-PLEX Human CTACK Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of CTACK in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can perform up to 80 tests across two plates The lower limit of detection is 0 05 pg/mL The kit includes a 96-well MULTI-ARRAY Human CTACK Calibrator and all necessary reagents and plates for the assay
Catalog No. 50-000-79829 Supplier Meso Scale Discovery Supplier No. K151VDK2
The Meso Scale Discovery U-PLEX Human CTACK Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of CTACK in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can perform up to 80 tests across two plates The lower limit of detection is 0 05 pg/mL The kit includes a 96-well MULTI-ARRAY Human CTACK Calibrator and all necessary reagents and plates for the assay