Learn More
Meso Scale Discovery U-PLEX Human CTACK ASY
SDP
Catalog No. 5000081434
Encompass
The Meso Scale Discovery U-PLEX Human CTACK Assay is a multiplex immunoassay kit that allows simultaneous measurement of CTACK and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers It includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator This assay enables quantification of CTACK levels which may have research and diagnostic utility
Catalog No. 50-000-81434 Supplier Meso Scale Discovery Supplier No. K151VDK1
The Meso Scale Discovery U-PLEX Human CTACK Assay is a multiplex immunoassay kit that allows simultaneous measurement of CTACK and other biomarkers in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers It includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator This assay enables quantification of CTACK levels which may have research and diagnostic utility