Learn More
Meso Scale Discovery U-PLEX Human CTACK ASY384-well
SDP
Catalog No. 5000081129
Encompass
The Meso Scale Discovery U-PLEX Human CTACK 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human cytokine CTACK in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of up to 80 samples The assay has a sensitivity of 0 08 pg/mL and requires just 12 5 L of sample per well The kit includes a 384-well MULTI-ARRAY Meso Scale Discovery plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator
Catalog No. 50-000-81129 Supplier Meso Scale Discovery Supplier No. K251VDK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CTACK 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human cytokine CTACK in cell culture supernatants serum plasma and other biological matrices It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of up to 80 samples The assay has a sensitivity of 0 08 pg/mL and requires just 12 5 L of sample per well The kit includes a 384-well MULTI-ARRAY Meso Scale Discovery plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator