Learn More
Meso Scale Discovery U-PLEX Human CTACK ASYQP
SDP
Catalog No. 5000081435
Encompass
The U-PLEX Human CTACK Assay (K151VDK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of CTACK (CCL27) chemokine levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing of multiple samples The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in research applications studying immune responses and inflammatory processes
Catalog No. 50-000-81435 Supplier Meso Scale Discovery Supplier No. K151VDK22
The U-PLEX Human CTACK Assay (K151VDK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of CTACK (CCL27) chemokine levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing of multiple samples The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in research applications studying immune responses and inflammatory processes