Learn More
Meso Scale Discovery U-PLEX Human CTACK ASYQP
SDP
Catalog No. 5000082360
Encompass
The U-PLEX Human CTACK Assay (K151VDK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure CTACK (CCL27) levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantifying the chemokine CTACK which plays a role in skin-homing T-cell recruitment The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying inflammatory responses and immune cell migration
Catalog No. 50-000-82360 Supplier Meso Scale Discovery Supplier No. K151VDK21
The U-PLEX Human CTACK Assay (K151VDK) is a multiplex immunoassay kit from Meso Scale Discovery designed to measure CTACK (CCL27) levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity and specificity for quantifying the chemokine CTACK which plays a role in skin-homing T-cell recruitment The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying inflammatory responses and immune cell migration