Learn More
Meso Scale Discovery U-PLEX Human CTLA-4 ASY
SDP
Catalog No. 5000078260
Encompass
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay for measuring CTLA-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption Requiring 50 L of sample it is compatible with MESO QuickPlex SQ 120 instruments The kit includes a plate with capture antibodies detection antibodies and calibrators for a standard curve enabling quantitative CTLA-4 measurement in about 3 5 hours
Catalog No. 50-000-78260 Supplier Meso Scale Discovery Supplier No. K151R7K2
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay for measuring CTLA-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample consumption Requiring 50 L of sample it is compatible with MESO QuickPlex SQ 120 instruments The kit includes a plate with capture antibodies detection antibodies and calibrators for a standard curve enabling quantitative CTLA-4 measurement in about 3 5 hours