Learn More
Meso Scale Discovery U-PLEX Human CTLA-4 ASY
SDP
Catalog No. 5000082195
Encompass
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay for measuring CTLA-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 series The kit includes a plate with capture antibodies detection antibodies and calibrators for a standard curve allowing quantitative CTLA-4 measurement in about 3 5 hours
Catalog No. 50-000-82195 Supplier Meso Scale Discovery Supplier No. K151R7K1
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay for measuring CTLA-4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 series The kit includes a plate with capture antibodies detection antibodies and calibrators for a standard curve allowing quantitative CTLA-4 measurement in about 3 5 hours