Learn More
Meso Scale Discovery U-PLEX Human CTLA-4 ASYQP
SDP
Catalog No. 5000079881
Encompass
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay that measures CTLA-4 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 MESO SECTOR S 600 and MESO SECTOR S 6 Each kit contains a plate coated with capture antibodies detection antibodies and calibrators to generate a standard curve
Catalog No. 50-000-79881 Supplier Meso Scale Discovery Supplier No. K151R7K21
The Meso Scale Discovery U-PLEX Human CTLA-4 Assay Kit is a multiplex immunoassay that measures CTLA-4 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 MESO SECTOR S 600 and MESO SECTOR S 6 Each kit contains a plate coated with capture antibodies detection antibodies and calibrators to generate a standard curve