Learn More
Meso Scale Discovery U-PLEX Human Cystatin C ASY
SDP
Catalog No. 5000079956
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit quantifies Cystatin C in human serum plasma and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the QuickPlex SQ 120 It detects from 0 08 ng/mL to 2500 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent offering a simple rapid and sensitive method for measuring Cystatin C in human samples
Catalog No. 50-000-79956 Supplier Meso Scale Discovery Supplier No. K151M9K2
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit quantifies Cystatin C in human serum plasma and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the QuickPlex SQ 120 It detects from 0 08 ng/mL to 2500 ng/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent offering a simple rapid and sensitive method for measuring Cystatin C in human samples