Learn More
Meso Scale Discovery U-PLEX Human Cystatin C ASY
SDP
Catalog No. 5000081180
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Cystatin C in human serum plasma and urine It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive accurate and rapid quantification of Cystatin C from small sample volumes This assay can aid in the diagnosis and monitoring of kidney function and related diseases It has a measuring range of 50-10 000 pg/mL and exhibits minimal cross-reactivity to related proteins The kit includes a 96-well plate detection antibody calibrator and assay diluent valid for 50 tests
Catalog No. 50-000-81180 Supplier Meso Scale Discovery Supplier No. K151M9K1
The Meso Scale Discovery U-PLEX Human Cystatin C Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Cystatin C in human serum plasma and urine It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive accurate and rapid quantification of Cystatin C from small sample volumes This assay can aid in the diagnosis and monitoring of kidney function and related diseases It has a measuring range of 50-10 000 pg/mL and exhibits minimal cross-reactivity to related proteins The kit includes a 96-well plate detection antibody calibrator and assay diluent valid for 50 tests