Learn More
Meso Scale Discovery U-PLEX Human Cystatin C ASYQP
SDP
Catalog No. 5000079957
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit measures Cystatin C in human serum plasma and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results similar to ELISA The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a simple rapid high-throughput method to quantify Cystatin C levels
Catalog No. 50-000-79957 Supplier Meso Scale Discovery Supplier No. K151M9K21
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit measures Cystatin C in human serum plasma and other fluids This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results similar to ELISA The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a simple rapid high-throughput method to quantify Cystatin C levels