Learn More
Meso Scale Discovery U-PLEX Human Cystatin C ASYQP
SDP
Catalog No. 5000079295
Encompass
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit is a multiplex immunoassay that measures human Cystatin C in serum plasma and other biological fluids This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrator and assay diluent This quantitative assay has a typical sensitivity of 0 3 ng/mL and requires 25 L of sample The U-PLEX Human Cystatin C Assay provides a simple rapid and sensitive method to measure Cystatin C levels
Catalog No. 50-000-79295 Supplier Meso Scale Discovery Supplier No. K151M9K22
The Meso Scale Discovery U-PLEX Human Cystatin C Assay Kit is a multiplex immunoassay that measures human Cystatin C in serum plasma and other biological fluids This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrator and assay diluent This quantitative assay has a typical sensitivity of 0 3 ng/mL and requires 25 L of sample The U-PLEX Human Cystatin C Assay provides a simple rapid and sensitive method to measure Cystatin C levels