Learn More
Meso Scale Discovery U-PLEX Human DPPIV ASY 25 PL
SDP
Catalog No. 5000081608
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of DPP4 and other biomarkers in human serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of 10 000 ng/mL for DPP4
Catalog No. 50-000-81608 Supplier Meso Scale Discovery Supplier No. K151D9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human DPP4 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of DPP4 and other biomarkers in human serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of 10 000 ng/mL for DPP4