Learn More
Meso Scale Discovery U-PLEX Human DPPIV ASY
SDP
Catalog No. 5000080165
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 Assay Kit is a multiplex immunoassay that measures Dipeptidyl Peptidase 4 (DPP4) levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator and controls Sample incubation time is 2 hours and total assay time is 3 5 hours The assay is designed for the quantitative measurement of DPP4 levels for life science research applications
Catalog No. 50-000-80165 Supplier Meso Scale Discovery Supplier No. K151D9K1
The Meso Scale Discovery U-PLEX Human DPP4 Assay Kit is a multiplex immunoassay that measures Dipeptidyl Peptidase 4 (DPP4) levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator and controls Sample incubation time is 2 hours and total assay time is 3 5 hours The assay is designed for the quantitative measurement of DPP4 levels for life science research applications