Learn More
Meso Scale Discovery U-PLEX Human DPPIV ASY
SDP
Catalog No. 5000080166
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 Assay is a multiplex immunoassay panel that measures Dipeptidyl Peptidase 4 (DPP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on MSD s QuickPlex SQ 120 instrument The assay has high sensitivity and wide dynamic range It requires a low sample volume of 50 L The total assay time is approximately 3 5 hours This product provides a fast simple and robust way to measure DPP4 levels across multiple samples
Catalog No. 50-000-80166 Supplier Meso Scale Discovery Supplier No. K151D9K2
The Meso Scale Discovery U-PLEX Human DPP4 Assay is a multiplex immunoassay panel that measures Dipeptidyl Peptidase 4 (DPP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on MSD s QuickPlex SQ 120 instrument The assay has high sensitivity and wide dynamic range It requires a low sample volume of 50 L The total assay time is approximately 3 5 hours This product provides a fast simple and robust way to measure DPP4 levels across multiple samples