Learn More
Meso Scale Discovery U-PLEX Human DPPIV ASY384-well
SDP
Catalog No. 5000080280
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of DPP4 in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a simple no-wash protocol with high sensitivity and broad dynamic range The kit includes a 384-well plate coated with capture antibody detection antibody labeled with MSD GOLD SULFO-TAG calibrators and diluent The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL
Catalog No. 50-000-80280 Supplier Meso Scale Discovery Supplier No. K251D9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human DPP4 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of DPP4 in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a simple no-wash protocol with high sensitivity and broad dynamic range The kit includes a 384-well plate coated with capture antibody detection antibody labeled with MSD GOLD SULFO-TAG calibrators and diluent The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL