Learn More
Meso Scale Discovery U-PLEX Human DPPIV ASYQP
SDP
Catalog No. 5000081607
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 Assay Kit is a multiplex immunoassay for measuring Dipeptidyl Peptidase 4 (DPP4) in human serum plasma and cell culture supernatant It uses electrochemiluminescent detection for high sensitivity and rapid results The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and diluent Sensitivity is 0 3 ng/mL with an upper quantification limit of 10 000 ng/mL requiring 25 L of sample It is compatible with MSD instruments offering a sensitive and reproducible method for DPP4 quantification
Catalog No. 50-000-81607 Supplier Meso Scale Discovery Supplier No. K151D9K22
The Meso Scale Discovery U-PLEX Human DPP4 Assay Kit is a multiplex immunoassay for measuring Dipeptidyl Peptidase 4 (DPP4) in human serum plasma and cell culture supernatant It uses electrochemiluminescent detection for high sensitivity and rapid results The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and diluent Sensitivity is 0 3 ng/mL with an upper quantification limit of 10 000 ng/mL requiring 25 L of sample It is compatible with MSD instruments offering a sensitive and reproducible method for DPP4 quantification