Learn More
Meso Scale Discovery U-PLEX Human DPPIV ASYQP
SDP
Catalog No. 5000081977
Encompass
The Meso Scale Discovery U-PLEX Human DPP4 Assay is an electrochemiluminescent immunoassay for measuring DPP4 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the Meso QuickPlex SQ 120 The assay detects 50-10 000 pg/mL with no cross-reactivity using 50 L samples The kit includes a 96-well plate with capture antibody detection antibody with ruthenium calibrators and reagents offering a reproducible high-throughput method for quantifying DPP4
Catalog No. 50-000-81977 Supplier Meso Scale Discovery Supplier No. K151D9K21
The Meso Scale Discovery U-PLEX Human DPP4 Assay is an electrochemiluminescent immunoassay for measuring DPP4 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the Meso QuickPlex SQ 120 The assay detects 50-10 000 pg/mL with no cross-reactivity using 50 L samples The kit includes a 96-well plate with capture antibody detection antibody with ruthenium calibrators and reagents offering a reproducible high-throughput method for quantifying DPP4