Learn More
Meso Scale Discovery U-PLEX Human E-Selectin ASY
SDP
Catalog No. 5000081575
Encompass
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit for measuring E-Selectin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The assay requires 50 L or less sample volume and is compatible with MSD instruments It includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables sensitive reproducible quantification of E-Selectin a key biomarker of endothelial cell activation
Catalog No. 50-000-81575 Supplier Meso Scale Discovery Supplier No. K151AGVK1
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit for measuring E-Selectin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The assay requires 50 L or less sample volume and is compatible with MSD instruments It includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables sensitive reproducible quantification of E-Selectin a key biomarker of endothelial cell activation