Learn More
Meso Scale Discovery U-PLEX Human E-Selectin ASYQP
SDP
Catalog No. 5000079048
Encompass
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 0 05 ng/mL
Catalog No. 50-000-79048 Supplier Meso Scale Discovery Supplier No. K151AGVK22
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of E-Selectin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates The lower limit of detection is 0 05 ng/mL