Learn More
Meso Scale Discovery U-PLEX Human E-Selectin ASYQP
SDP
Catalog No. 5000080595
Encompass
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit for measuring E-Selectin in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and precision Requiring 50 L or less sample volume it can perform up to 80 tests on two plates with a detection limit of 0 03 ng/mL Results are available in 3 5 hours offering a fast sensitive and reproducible method for quantifying E-Selectin levels in various human samples
Catalog No. 50-000-80595 Supplier Meso Scale Discovery Supplier No. K151AGVK21
The Meso Scale Discovery U-PLEX Human E-Selectin Assay is a multiplex immunoassay kit for measuring E-Selectin in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and precision Requiring 50 L or less sample volume it can perform up to 80 tests on two plates with a detection limit of 0 03 ng/mL Results are available in 3 5 hours offering a fast sensitive and reproducible method for quantifying E-Selectin levels in various human samples