Learn More
Meso Scale Discovery U-PLEX Human ENA-78 ASYQP
SDP
Catalog No. 5000079700
Encompass
The MSD U-PLEX Human ENA-78 Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of ENA-78 (epithelial neutrophil-activating protein-78) in human biological samples This singleplex assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements for research applications studying inflammatory responses and chemokine signaling pathways
Catalog No. 50-000-79700 Supplier Meso Scale Discovery Supplier No. K151VEK21
The MSD U-PLEX Human ENA-78 Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of ENA-78 (epithelial neutrophil-activating protein-78) in human biological samples This singleplex assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements for research applications studying inflammatory responses and chemokine signaling pathways