Learn More
Meso Scale Discovery U-PLEX Human Eotaxin-2 ASY
SDP
Catalog No. 5000079624
Encompass
The Meso Scale Discovery U-PLEX Human Eotaxin-2 Assay Kit is designed for the quantitative measurement of human Eotaxin-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary U-PLEX technology is sensitive reproducible and has a broad dynamic range of 15 6 - 1000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized Eotaxin-2 calibrator The assay takes 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-79624 Supplier Meso Scale Discovery Supplier No. K151XQK1
The Meso Scale Discovery U-PLEX Human Eotaxin-2 Assay Kit is designed for the quantitative measurement of human Eotaxin-2 in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary U-PLEX technology is sensitive reproducible and has a broad dynamic range of 15 6 - 1000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized Eotaxin-2 calibrator The assay takes 3 5 hours to complete after a 2 hour incubation